Please login to your online account to display your discounted pricing
Kimble™ Kontes™ Wide Mouth Heavy Duty Class A Volumetric Flasks with Screw Cap
Designed to ASTM for class A volumetric standards and are calibrated to contain
$126.55 - $126.55
Specifications
Material Borosilicate Glass
Mouth Style Wide
View More Specs
Products
Catalog Number Mfr. No. Capacity (Metric) Length (Metric) Tolerance Price Quantity    
K6231000050
kimble™
6231000050
50mL 140mm ±0.05mL Each for $126.55
Description & Specifications
Specifications
Material Borosilicate Glass
Mouth Style Wide
  • Heavy wall design for added durability
  • Wide mouth with screw thread opening for easy access when using a pipet or adding solid material
  • Mixing bulb is useful in dissolving tablets and other solids
  • Screw thread feature allows safe and secure storage
  • Cap is chemically resistant and provides an air-tight seal
  • Flask can be used in high temperature applications as high as 180°C