Please login to your online account to display your discounted pricing
Kimble™ Kontes™ Vacuum Filter Holders:: Ultra-ware Filtration Adapter::Replacement Parts
Manufacturer: kimble™  4101704534
 View more versions of this product
Catalog No. K410170-4534
  • / Each
Description & Specifications
Specifications
Description Cap
For Use With (Equipment) Kimble-Chase Kontes Ultra-ware Filtration Adapter
Material PBT
Diameter (Metric) Inner 34mm