DWK Life Sciences Kimble™ Kontes™ Universal Inlet with Hose Connection
Highly adaptable for vacuum seal
Supplier: DWK Life Sciences 1798502224
Description
This universal connector is designed to adapt a variety of tubes, condensers and filters in 14standard taper/20 and 24standard taper/25-24standard taper/40 glassware. Diameter variations of 1 to 2mm can be accommodated since the inert O-ring is compressed by the polypropylene cap to effect a vacuum tight seal.
- Serrated side port can be used for vacuum connection or for introduction of gases
- Polypropylene compression cap is suitable for use to 150°C
- Provides a combination inlet and vent tube on a single neck vessel
- Converts plain stem type funnels for vacuum filtration
- Extends gas inlet or gas dispersion tube into a flask to function as an impinger
- Serves as a pressure relief vent when used with additional funnels
- Can be used to construct gas washing bottles
Specifications
Hose Inlet Adapter | |
PTFE | |
9.3mm | |
Polypropylene compression cap |