Please login to your online account to display your discounted pricing

Kimble™ Kontes™ ULTRA-WARE™ Heavy-Wall Filter Flasks


Heavy-wall borosilicate glass provides mechanical strength for vacuum filtration

Manufacturer: kimble™  9537601002

 View more versions of this product

Catalog No. K9537601002

  • / Each
 

Description & Specifications

Specifications

Capacity (Metric) 1000mL
Closure Size 8
Diameter (English) Inner Tubing 0.37 in.
Diameter (Metric) Inner Tubing 0.93cm
Material Borosilicate Glass

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.