Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Two Neck Round Bottom Heavy Wall Distilling Flasks


Manufacturer: kimble™  2949900100

 View more versions of this product

Catalog No. K294990-0100

  • / Each
 

Description & Specifications

Specifications

Neck Style Angled
Diameter (Metric) Outer 60mm
Material Borosilicate Glass
Standard Taper Joint Side 14/20
Capacity (Metric) 100mL
No. of Positions 2.0
Standard Taper Joint 14/20 Center, 14/20 side
Diameter (English) Outer 2.3 in.
Bottom Shape Round
Includes Standard taper outer joint