Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Two Neck Round Bottom Heavy Wall Distilling Flasks


Manufacturer: kimble™  2949900010

 View more versions of this product

Catalog No. K294990-0010

  • / Each
 

Description & Specifications

Specifications

Bottom Shape Round
Capacity (Metric) 10mL
Diameter (English) Outer 1.2 in.
Diameter (Metric) Outer 31mm
Includes Standard taper outer joint
Neck Style Angled
No. of Positions 2.0
Standard Taper Joint Side 14/20
Standard Taper Joint 14/20 Center, 14/20 side
Material Borosilicate Glass