Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Tubes For Dounce All-Glass Tissue Grinders


For Dounce All-Glass tissue grinders

Manufacturer: kimble™  8853030007

 View more versions of this product

Catalog No. K885303-0007

  • / Each
 

Description & Specifications

Specifications

For Use With (Equipment) Dounce all Glass tissue grinders
Capacity (Metric) 7mL
Capacity (English) 0.23 oz.
Product Type Tissue grinder tube
Capacity 0.23 oz., 7mL