Kimble™ Kontes™ Tubes for Duall™ All-Glass Tissue Grinders
Improves grinding efficiency
Manufacturer:  Kimble™ 8854620022
 View more versions of this product
Catalog No. K885462-0022
Specifications
Description
Tube; Kimble-Chase Kontes; Component part; Size 22; For Duall all-glass tissue grinders; With plastic safety coating
  • Tubes sizes from 21 to 25
  • Available with or without coating
Description & Specifications
Specifications
Duall™ all Glass tissue grinders
22
With plastic safety coating
Improves grinding efficiency
Plastic
Tissue grinder tube