Kimble™ Kontes™ Tenbroeck Tissue Grinder
Hand or motor operation
$142.71 - $319.00
Specifications
Clearance (English) 0.004 to 0.006 in.
Clearance (Metric) 0.1 to 0.15mm
Product Type Tenbroeck tissue grinder
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity Pestle Opening Stopper Number Material Price Quantity  
Catalog Number Mfr. No. Capacity Pestle Opening Stopper Number Material Price Quantity  
06-421-40 Kimble™
K8850000040
1.3 oz., 40mL 3 Glass Each for $319.00
K885000-0007 Kimble™
8850000007
0.23 oz., 7mL 0 Ground Glass Case for $147.76
K885000-0002 Kimble™
8850000002
0.06 oz., 2mL 0 Ground Glass Each for $142.71
K885000-0015 Kimble™
8850000015
0.5 oz., 15mL 1 Ground Glass Each for $164.91
Specifications
Description
  • All-glass construction
  • Pestle is tooled for an appropriate sized single hole rubber stopper (sold separately)
  • Can be motor driven at slow speeds by inserting the metal rod into the motor chuck
  • Clearance between pestles and tubes is 0.1 to 0.15mm
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
0.004 to 0.006 in.
0.1 to 0.15mm
Tenbroeck tissue grinder