Please login to your online account to display your discounted pricing
Kimble™ Kontes™ Straight with Tip-O-Ring PTFE Plug HI-VAC Valves
For vacuum applications up to 5 x 10-7 torr
$98.89 - $98.89
Specifications
Product Type Straight valve
For Use With (Application) For vacuum appliCatIons up to 5 x 10-7 torr
Material Borosilicate Glass
View More Specs
 Alert
Polyacetal knob is not recommended for use over 110°C.
Products
Catalog Number Mfr. No. Bore Size (English) Diameter (English) Outer Stem Range Shaft O-Ring Size Price Quantity    
K826500-0004
kimble™
8265000004
0 to 0.15 in. 0.35 in. 0 to 4mm 010 Each for $98.89
Description & Specifications
Specifications
Product Type Straight valve
For Use With (Application) For vacuum appliCatIons up to 5 x 10-7 torr
Material Borosilicate Glass
  • Comes with precision external thread for fine control
  • Non-rotating shaft with primary seal made with Viton™ o-ring on tapered seat
  • Full open position at 1-1/2 turns
  • Used as bleed at up to half turn
  • PTFE shaft can be used continuously at system temperatures of 230°C or intermittently at 275°C