Please login to your online account to display your discounted pricing
Kimble™ Kontes™ Straight with Tip-O-Ring Glass Plugs
Uses glass plug for applications which do not require “out-gassing” plug material
Manufacturer: kimble™  8264510004
 View more versions of this product
 Alert
Polyacetal knob is not recommended for use over 110°C.
Catalog No. K826451-0004
  • / Each
Description & Specifications
Specifications
Material Glass
Plug Stopcock No 4
Product Type Replacement plug