Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Spargers for Tekmar™ Equipment


For use with Tekmar 2000 and 3000 Purge and Trap Concentrators and with 2016/2032 Autosamplers

Manufacturer: kimble™  5911002625*

 View more versions of this product

Catalog No. K591100-2625

  • / Each
 

Description & Specifications

Specifications

Capacity (English) 0.84 oz.
Capacity 0.84 oz., 25mL
Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
For Use With (Application) Potable water and wastewater samples
For Use With (Equipment) Teledyne Tekmar™ 2000 and 3000 purge and trap concentrators and with the 2016 and 2032 autosamplers
Material Borosilicate Glass
Shape U
Capacity (Metric) 25mL