Please login to your online account to display your discounted pricing
Kimble™ Kontes™ Soxhlet Extraction Apparatus, Micro
$150.02 - $549.00
Specifications
Includes Reflux condenser, extractor body and round bottom flask
View More Specs
Includes:  Complete apparatus includes reflux condenser, extractor body, and round-bottom flask
Requires:  Requires 10 × 50mm Glass Extraction Thimble
Products
Catalog Number Mfr. No. Type Price Quantity    
K283250-0000
kimble™
2832500000
Soxhlet Condenser Each for $150.02
K292010-0000
kimble™
2920100000
Extraction Apparatus Each for $549.00
K292011-0000
kimble™
2920110000
Extractor Body Each for $380.00
Description & Specifications
Specifications
Includes Reflux condenser, extractor body and round bottom flask