Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Round Bottom Three Neck Heavy Wall Distilling Flasks


With outer joints

Manufacturer: kimble™  6060002924

 View more versions of this product

Catalog No. K606000-2924

  • / Each
 

Description & Specifications

Specifications

Capacity (Metric) 5000mL
No. of Positions 3.0
Diameter (Metric) Outer 221mm
Neck Style Vertical
Includes Standard taper outer joint
Material Borosilicate Glass
Bottom Shape Round
Standard Taper Joint Side 24/40
Standard Taper Joint 45/50 Center, 24/40 side
Diameter (English) Outer 8.7 in.