missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kontes™ Disposable Microtubes

Manufacturer:  Kimble Chase Life Science 7495301500

 View more versions of this product

Catalog No. K749530-1500


 

Specifications

Description

Microtubes; Kimble-Chase Kontes; Polypropylene; Autoclavable; Note: Pellet pestle sold separately; 15mL; Natural; Pack of 25

  • Autoclavable polypropylene. Available colors: blue, natural.

Description & Specifications

Specifications

0.5 oz., 15mL
Pellet Pestles™
Yes
15mL
Natural
Polypropylene
0.5 oz.