Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Reagent Sprayers with Screw Thread Ground Joint


Provide uniform spray mist of visualization reagents for TLC and paper chromatography; use with all standard TLC reagents and most corrosive liquids

Manufacturer: kimble™  4225300050

 View more versions of this product

Catalog No. K422530-0050

  • / Each
 

Description & Specifications

Specifications

Product Type TLC reagent sprayer
Connections Screw thread ground joint, connecting cap, loosening ring
Operating Pressure 3 to 5psi
Quantity 1 Case
For Use With (Application) To provide uniform spray mist of visualization reagents for TLC and paper chromatography
Diameter (English) Outer 1.96 in.
Standard Taper Joint 19/22
Capacity 1.69 oz., 50mL
Height (English) 6.85 in.
Height (Metric) 17.4cm
Diameter (Metric) Outer 5cm
Autoclavable Yes
Material Glass
Capacity (Metric) 50mL
For Use With (Equipment) Standard TLC reagents and most corrosive liquids
Capacity (English) 1.69 oz.