missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimble™ Kontes™ PTFE-Plugged Columns

With or without fritted disc

Manufacturer:  Kimble™ 4205300225

 View more versions of this product

Catalog No. K420530-0225


 

Specifications Description & Specifications

Specifications

Analytical separation
80mL
Glass
15mm
Plain Chromatography Column
500mm
Glass Wool
2mm Bore PTFE stopcock plug