missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimble™ Kontes™ PTFE-Plugged Columns

With or without fritted disc

Manufacturer:  Kimble™ 4205300253

 View more versions of this product

Catalog No. K420530-0253


 

Specifications Description & Specifications

Specifications

Analytical separation
300mL
Glass
30mm
Plain Chromatography Column
500mm
Glass Wool
2mm Bore PTFE stopcock plug