Please login to your online account to display your discounted pricing

Kimble™ Kontes™ PTFE-Plugged Columns


With or without fritted disc

Manufacturer: kimble™  4205300242

 View more versions of this product

Catalog No. K420530-0242

  • / Each
 

Description & Specifications

Specifications

Includes 2mm Bore PTFE stopcock plug
Column Capacity 130mL
Length (Metric) 350mm
Diameter (Metric) Inner 22mm
Type Plain Chromatography Column
Material Glass
Support Material Glass Wool
For Use With Analytical separation