Kimble™ Kontes™ Potter-Elvehjem Tissue Grinders
With serrated PTFE pestle and graduated tube
$166.00 - $266.09
Specifications
Product Type Pestle and graduated tube
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity Graduations Price Quantity  
Catalog Number Mfr. No. Capacity Graduations Price Quantity  
K885510-0023 Kimble™
8855100023
1.85 oz., 55mL 6 to 50mL Each for $266.09
K885510-0020 Kimble™
8855100020
0.13 oz., 4mL 0.5 to 4mL Each for $222.15
K885510-0021 Kimble™
8855100021
0.33 oz., 10mL 1 to 9mL Each for $166.00
K885510-0022 Kimble™
8855100022
1.01 oz., 30mL 2 to 25mL Each for $225.44
Specifications
Description
  • Radial serrations on the lower portion of PTFE pestle deliver homogenate into cylindrical portion of tube
  • Stainless-steel pestle shaft
  • Graduations fused into the glass tube
  • Tubes and pestles are interchangeable
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Pestle and graduated tube