Kimble™ Kontes™ Polypropylene Closure with Ultrasonically Welded PTFE Liner
For use with screw-thread culture and centrifuge tubes
Manufacturer:  Kimble™ K7380824400
 View more versions of this product
Catalog No. 03-340-65
Specifications Description & Specifications
Specifications
Yes
PTFE, Rubber
For repeated autoclaving
24-400
PTFE
Polypropylene
Screw Cap
EPA water analysis vials, screw thread culture and centrifuge tubes
24-400