missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Pestles For DUALL™ All-Glass Tissue Grinders

For DUALL All-Glass tissue grinders

$91.87 - $97.18

Specifications

Product Type Tissue grinder pestle
For Use With (Equipment) Duall™ all Glass tissue grinders
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
K885451-0020 Kimble™
8854510020
20 Each for $97.18
K885451-0021 Kimble™
8854510021
21 Each for $97.18
K885451-0022 Kimble™
8854510022
22 Each for $91.87
K885451-0023 Kimble™
8854510023
23 Each for $94.31
Specifications

Description

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

Tissue grinder pestle
Duall™ all Glass tissue grinders