Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Pestles For Dounce All-Glass Tissue Grinders


All-glass component parts

Manufacturer: kimble™  8853010002

 View more versions of this product

Catalog No. K885301-0002

  • / Each
 

Description & Specifications

Specifications

Capacity (Metric) 2mL
For Use With (Equipment) Dounce tissue grinder
Capacity (English) 0.06 oz.
Capacity 0.06 oz., 2mL
Product Type Large clearance pestle