Please login to your online account to display your discounted pricing
Kimble™ Kontes™ Packing Reservoirs for Chromaflex Columns
Manufacturer: kimble™  4208370040
 View more versions of this product
Catalog No. K420837-0040
  • / Each
Description & Specifications
Specifications
Diameter 4.8cm
Quantity 2000mL
Packaging End fitting has 1/4-28 threads for easy connection to pumps