Please login to your online account to display your discounted pricing
Kimble™ Kontes™ Packing Reservoirs for Chromaflex Columns
Manufacturer: kimble™  K4208370020
 View more versions of this product
Catalog No. K420837-0020
  • / Each
Description & Specifications
Specifications
Packaging End fitting has 1/4-28 threads for easy connection to pumps
Quantity 500mL
Diameter 2.5cm