Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Microscale Organic Chemistry LABSETS™ and Kits Components


Components for microscale organic chemistry LABSETS

Manufacturer: kimble™  7480091005

 View more versions of this product

Catalog No. K748009-1005

  • / Each
 

Description & Specifications

Specifications

Capacity (English) 0.16 oz.
Capacity (Metric) 5mL
Capacity 0.16 oz., 5mL
For Use With (Application) Microscale organic chemistry
For Use With (Equipment) Component part of Williamson standard organic chemistry labsets and kits
Quantity Ea.
Product Type Long Neck Flask