Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Microscale Organic Chemistry LABSETS™ and Kits Components


Components for microscale organic chemistry LABSETS

Manufacturer: kimble™  7480031000*

 View more versions of this product

Catalog No. K748003-1000

  • / Case of 5 Each
 

Description & Specifications

Specifications

For Use With (Application) Microscale organic chemistry
Quantity 5/Pk.
Product Type Thermometer Adapter