Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Microscale Organic Chemistry LABSETS™ and Kits Components


Components for microscale organic chemistry LABSETS

Manufacturer: kimble™  7480020000

 View more versions of this product

Catalog No. K748002-0000

  • / Pack of 10 Each
 

Description & Specifications

Specifications

For Use With (Equipment) For Kontes microscale organic chemistry labsets
Quantity 10/Pk.
Product Type Filter Adapter
Includes Pluro stopper
For Use With (Application) Microscale organic chemistry
Standard Taper Joint 14/20