Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Microscale Organic Chemistry LABSETS™ and Kits Components


Components for microscale organic chemistry LABSETS

Manufacturer: kimble™  7480010003

 View more versions of this product

Catalog No. 11-304-16

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Microscale organic chemistry
For Use With (Equipment) For Microflex™ organic chemistry kits
Quantity Ea.
Product Type Foam Insert Set