Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Microscale Organic Chemistry LABSETS™ and Kits Components


Components for microscale organic chemistry LABSETS

Manufacturer: kimble™  7480051001

 View more versions of this product

Catalog No. K748005-1001

  • / Case of 5 Each
 

Description & Specifications

Specifications

For Use With (Application) Microscale organic chemistry
Quantity 5/Pk.
Product Type Connector, Component Part

  • Microflex Kits feature special chaste connectors and specially tooled glassware components, providing great versatility
  • They fit easily into lab bench drawers