DWK Life Sciences Kimble™ Kontes™ Micro-Vial Graduated Screw Thread Vials
With solid-top closure and PTFE-faced white rubber liner
Manufacturer:  DWK Life Sciences 607071
 View more versions of this product
Catalog No. 06-100-1P
Description
Description
  • Accuform™
  • KG-33 borosilicate glass
  • Extra thick clear glass walls
Specifications
Specifications
Amber
Borosilicate Glass
Yes
PTFE/Rubber
13-425
49 mm
14 mm
1 mL
1.92 in.
Tapered
Screw Cap
12 Case