missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Kimble™ Kontes™ Macroscale Organic Chemistry Kit

Designed to address the experiments detailed in the Williamson text Macroscale Organic Experiments published by D.C. Heath and Company, Lexington, Massachusetts

Supplier:  DWK Life Sciences 2693300000

Catalog No. K269330-0000



Includes:
  • 15mL Conical flask
  • Distillation adapters, connecting, thermometer, BEVEL-SEAL™
  • Distillation adapters, Bent, 105°, with vacuum take-off
  • Plain distillation columns
  • Recovery flasks
  • 125mL PTFE separatory funnel
  • Nylon Body/HDPE plug
  • Chromatography Column
  • Funnel
  • Luer tip
  • 20 microns polyethylene disc
  • Polyacetal standard taper clamp
  • Filter adapter, Pluro stopper
  • 125mL Filter flask,
  • Hirsch funnel
  • standard taper Joint 14/20
  • Distillation adapters, Claisen
  • West Condensers, medium length standard taper joints, jacketed

Description

Description

  • Students use the separatory funnel for extractions and for liquid addition in reaction assemblies
  • Classical atmospheric, steam, fractional, and vacuum distillations are possible with kit components
  • Minimal set up time
  • Smaller quantities afford a greater degree of safety, improve air quality in the lab
  • standard taper Joint: 14/20

Specifications

Specifications

Macroscale Organic Chemistry Kit
For extractions and for liquid addition in reaction assemblies
Borosilicate Glass
USP Type I and ASTM E438 Type I, Class A
15mL conical flask, distillation adapters, connecting, thermometer, bevel-seal™, distillation adapters, bent, 105°, with vacuum take-off, plain distillation columns, recovery flasks, 125mL PTFE separatory funnel
14/20
SDS
Documents

Documents

Promotions

Promotions