DWK Life Sciences Kimble™ Kontes™ KIMAX™ KimCote™ Heavy-Duty Solution Bottle (Carboy)
Plastic-coated for added safety
Manufacturer: DWK Life Sciences KC14950500
Description
- KimCote™ plastic-coating for added safety in the event of breakage
- Autoclavable
- Ultra-clear and extremely durable
- Resistant to many common laboratory chemicals
- Unique texture provides non-slip handling, wet or dry
5 gallon size is designed from Federal Specification DD-B-597
Specifications
Rubber | |
294 mm | |
502 mm | |
19,000 mL | |
Clear | |
Glass |
No. 12 | |
19.76 in. | |
642.4 oz., 19000mL | |
642.4 oz. | |
11.57 in. |