Please login to your online account to display your discounted pricing
Kimble™ Kontes™ Incubation Flask Stoppers and Center Well
$246.79 - $404.67
Specifications
Quantity 144/Pk.
View More Specs
Products
Catalog Number Mfr. No. Product Type Material Price Quantity    
K882310-0000
kimble™
8823100000
Stopper Rubber Pack of 144 Each for $404.67
K882311-0000
kimble chase life science
8823110000
Sleeved stopper Rubber Case of 144 Each for $246.79
K882320-0000
kimble™
8823200000
Center well Polypropylene Case of 144 Each for $279.38
Description & Specifications
Specifications
Quantity 144/Pk.
Flask Stoppers
  • Rubber stoppers for flask top and sidearm provide a secure double seal
  • Top stopper has off-center hole for insertion of center well
  • Smaller sidearm stopper has no hole, but can be punctured with a syringe needle
  • Both stoppers fit 10 and 25mL flasks
Center Well
  • Fits snugly into hole of top flask stopper for sample collection in a variety of metabolic applications
  • Made of chemical-resistant polypropylene
  • Disposable
  • Can be easily cut off for transfer of flask contents to liquid scintillation counting vial
  • Measures 10mm dia. × 70mm L
Top and sidearm stoppers are not interchangeable.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.