Please login to your online account to display your discounted pricing
Kimble™ Kontes™ HPLC Economy 3-Hole Ultra-Ware Cap Systems
With plastic coating that blocks virtually all UV light up to 385 nm, preventing photodegradation of light sensitive mobile phases
Manufacturer: kimble™  9539301003
 View more versions of this product
Catalog No. K953930-1003
  • / Each
Description & Specifications
Specifications
Capacity (Metric) 10L
Description Economy Three Hole Cap System with 10 Liter Reservoir
Includes TFE/Propylene o-ring and Polypropylene screw collar, 10L reservoir
No. per Case 1 Cs.
For Use With (Equipment) Bottle