Kimble™ Kontes™ HPLC Economy 3-Hole Ultra-Ware Cap Systems
With plastic coating that blocks virtually all UV light up to 385 nm, preventing photodegradation of light sensitive mobile phases
$497.97 - $1,196.59
Specifications
No. per Case 1 Cs.
For Use With (Equipment) Bottle
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Includes Capacity (Metric) Price Quantity  
K953930-1002 Kimble™
9539301002
TFE/Propylene o-ring and Polypropylene screw collar, 1L reservoir 1L Each for $497.97
K953930-1003 Kimble™
9539301003
TFE/Propylene o-ring and Polypropylene screw collar, 10L reservoir 10L Each for $1,196.59
K953930-2002 Kimble™
9539302002
TFE/Propylene o-ring and Polypropylene screw collar, 2L reservoir 2L Each for $557.36
K953930-5002 Kimble™
9539305002
TFE/Propylene o-ring and Polypropylene screw collar, 5L reservoir 5L Each for $955.05
Specifications
Description
  • Made of type I, Class A borosilicate glass with a plastic safety coating
  • Cap body is made of PTFE
  • Supplied with TFE/propylene o-ring and polypropylene screw collar
  • Connecting threads on top of cap use standard 1/4-28 flangeless fittings
  • Two ports have 1/8 in. through holes, third port has 1/16 in. through hole
Description & Specifications
Specifications
1 Cs.
Bottle