Please login to your online account to display your discounted pricing
Kimble™ Kontes™ Flow Adapters for Chromaflex™ Columns
$367.02 - $367.02
Specifications
Material Stainless Steel
View More Specs
Includes:  20μm porosity HDPE bed support, TFE/propylene O-ring seal, stainless-steel adjusting rod, PTFE body and 5 ft. (1.5m) of PTFE tubing.
Products
Catalog Number Mfr. No. Diameter (Metric) Inner Price Quantity    
K420836-0000
kimble™
4208360000
1cm Each for $367.02
Description & Specifications
Specifications
Material Stainless Steel
  • Delivers sample and buffer directly to top of gel bed
  • Ideal for concentration and pH gradients, reverse flow separations, and gravity flow separations that require a high level of resolution and reproducibility
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.