Please login to your online account to display your discounted pricing
Kimble™ Kontes™ Flip, Tear-Off Button Top Aluminum Seals, Color Coded
Designed to secure septa and stoppers to bottles and vials
Manufacturer: kimble™  K73844C13
 View more versions of this product
Catalog No. 03-340-334
  • / Case of 1000 Each
Description & Specifications
Specifications
Quantity 1000 Case
Type Seal
Material Closure Aluminum
Autoclavable Yes
Thread Size 13mm
Closure Material Aluminum
Closure Type Flip-up Button-top
Color Red