missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Kimble™ Kontes™ Flip-Off Button-Top Aluminum Seals, Color Coded

Two-piece, tamper-evident flip-off button top protects target area and allows easy access

Manufacturer:  DWK Life Sciences K73843B13

 View more versions of this product

Catalog No. 03-340-325



Description

Description

  • For use with 13 and 20mm serum finished bottles and vials
  • Designed to secure septa and stoppers to bottles and vials
  • Allows easy access with a flip of the thumb
  • Available with blue, red, green and white plastic buttons to help separate and identify samples

Specifications

Specifications

Yes
1000 Case
Flip-up Button-top Seal
Aluminum
13mm
Blue
Documents