missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimble™ Kontes™ FlexColumn™ Economy Columns

Ideal for use with FPLC™ systems, peristaltic pumps, or gravity flow to 15psi

Manufacturer:  Kimble™ 4204001550

 View more versions of this product

Catalog No. K420400-1550


 

Specifications Description & Specifications

Specifications

Economy Chromatography Column
2N NaOH or 100% Ethanol
14.5psi
15mm
Yes
20μm
89mL
HDPE
5/Cs.
1.77cm3
0 to 50°C
Male luer lock
Ion exchange, affinity, gel filtration and hydrophobic interaction media
Type I Class A Borosilicate Glass
500mm