Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Filtration Assemblies: Flasks With Inner Joint 40/35, for 90mm Glass Assemblies


Manufacturer: kimble™  9538280000

 View more versions of this product

Catalog No. K953828-0000

  • / Each
 

Description & Specifications

Specifications

Standard Taper Joint 40/35
Description Funnel for 47mm assemblies
For Use With (Equipment) Ultra-Ware 90mm all-glass Microfiltration Assembly
Capacity (Metric) 2000mL
Material Borosilicate Glass