Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Extraction Manifold Replacement Components


For extraction manifold

Manufacturer: kimble™  9538110000

 View more versions of this product

Catalog No. K953811-0000

  • / Pack of 25 Each
 

Description & Specifications

Specifications

Product Type PTFE Gasket
Length (Metric) 47mm
For Use With (Equipment) Solid Phase extraction manifold
Material PTFE
Size (Metric) Filter 4.7cm
Size (English) Filter 1.85 in.