Kimble™ Kontes™ Extraction Manifold Replacement Components
For extraction manifold.
Manufacturer:  Kimble™ 9711000090
 View more versions of this product
Catalog No. K971100-0090
Specifications
Description
Set, SPE Glassware; Kimble-Chase Kontes; For extraction manifold; 90mm Funnel, Base, Screen, 5 Gaskets, Clamp
  • For use with Kontes SPE Manifold Sets (971000-1047, -3047, or -6047)
  • Kontes SPE Glassware and Manifolds are specially designed as support systems for 47mm and 90mm dia. solid phase extraction disks to give you the highest possible recoveries of extracted analytes from your drinking water, surface water and waste water samples
Description & Specifications
Specifications
Solid Phase extraction manifold
90mm
SPE Glassware Set
90mm Funnel, support base, support screen, 5 PTFE gaskets and clamp
Glass