Please login to your online account to display your discounted pricing
Kimble™ Kontes™ Cyanide Distillation Apparatus with Sulfide Scrubber: Component Parts
Parts for use with Kimble-Chase Kontes Cyanide Distillation Apparatus with Sulfide Scrubber (cat. no. K479120-0000)
Manufacturer: kimble™  4791210000
 View more versions of this product
Catalog No. K479121-0000
  • / Each
Description & Specifications
Specifications
For Use With (Equipment) Kimble-Chase Kontes Cyanide Distillation Apparatus
Product Type Scrubber tube, standard taper 29/42 bottom, one standard taper 19/38 side and one spherical 18/7 side