DWK Life Sciences Kimble™ Kontes™ Replacement Pestle For All-Glass Dounce Tissue Grinders
Small and large clearance pestles for 15mL and 40mL Dounce Tissue Grinder.
$197.44 - $285.00
Specifications
For Use With (Equipment) Dounce all Glass tissue grinders
View More Specs
Products 3
Catalog Number Mfr. No. Capacity (Metric) Product Type Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Product Type Price Quantity  
K885302-0040
View Documents Promotion Details
DWK Life Sciences
8853020040
40 mL Small clearance pestle
Each for $285.00
K885301-0015
View Documents Promotion Details
DWK Life Sciences
8853010015
15 mL Large clearance pestle
Each for $197.44
K885302-0015
View Documents Promotion Details
DWK Life Sciences
8853020015
15 mL Small clearance pestle
Each for $207.53
Description
Description
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
Dounce all Glass tissue grinders
SDS
missing translation for 'documents'
missing translation for 'documents'