missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Column for Neutral Oils Content: Replacement Parts

For use with Kimble-Chase Kontes Column for Neutral Oils Content

Manufacturer:  Kimble™ 4271010000

 View more versions of this product

Catalog No. K427101-0000


 

Specifications Description & Specifications

Specifications

Low to medium pressure neutral oil and loss LC column
Solvent Reservoir
175mL