missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Kimble™ Kontes™ Column for Neutral Oils Content: Replacement Parts

For use with Kimble-Chase Kontes Column for Neutral Oils Content

$74.47 - $467.93

Specifications

For Use With (Equipment) Low to medium pressure neutral oil and loss LC column
View More Specs

Products 4
Catalog Number Mfr. No. Type Price Quantity  
Catalog Number Mfr. No. Type Price Quantity  
K427101-0000 View Documents DWK Life Sciences
4271010000
Solvent Reservoir Each for $265.93
 
K427103-0000 View Documents DWK Life Sciences
4271030000
Weighing Base Each for $74.47
 
K427102-0000 View Documents DWK Life Sciences
4271020000
Flask Each for $467.93
 
K427105-0000 View Documents DWK Life Sciences
4271050000
Liquid Chromatography (LC) Each for $292.32
 
Description

Description

For Column for Neutral Oils Content, see cat. no. K4271000000

Specifications

Specifications

Low to medium pressure neutral oil and loss LC column