Please login to your online account to display your discounted pricing
Kimble™ Kontes™ Chromatography Cylindrical TLC Developing Tanks
Lids for cylindrical glass tanks
Manufacturer: kimble™  4161701118
 View more versions of this product
Catalog No. K416170-1118
  • / Each
Description & Specifications
Specifications
Closure Material Polyethylene
For Use With (Application) Provides maximum vapor Phase saturation with a minimum amount of solvent
For Use With (Equipment) 1 x 3 in. Plate size
Holds 7.62 H x 2.54cm I.D. Plate
Material Polyethylene
Shape Cylindrical
Product Type Cylindrical TLC developing tank