missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Kimble™ Kontes™ Center Tear-Out Aluminum Seals, Color Coded
Designed to secure septa and stoppers to bottles and vials
$215.58 - $313.66
Catalog Number | Mfr. No. | Color | Thread Size | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Thread Size | Price | Quantity | ||||
03-340-312
|
DWK Life Sciences
73820B13 |
Blue | 13 mm |
Case of 1000 Each for $223.88
|
|
03-340-313
|
DWK Life Sciences
73820C13 |
Red | 13 mm |
Case of 1000 Each for $223.88
|
|
03-340-314
|
DWK Life Sciences
73820D13 |
Green | 13 mm |
Case of 1000 Each for $223.88
|
|
03-340-315
|
DWK Life Sciences
73820B20 |
Blue | 20 mm |
Case of 1000 Each for $215.58
|
|
03-340-317
|
DWK Life Sciences
73820D20 |
Green | 20 mm |
Case of 1000 Each for $313.66
|
|
03-340-316
|
DWK Life Sciences
73820C20 |
Red | 20 mm |
Case of 1000 Each for $215.58
|
|
Description
- Tamper-evident center tear-out feature protects target area
- For use with 13 and 20mm serum finished bottles and vials
- Available in blue, red, and green color to help separate and identify samples
Specifications
Tear-out | |
Aluminum | |
1000 Case |