missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Kimble™ Kontes™ Center Tear-Off Aluminum Seals, Color Coded
Tamper-evident center tear-off feature protects target area
$164.06 - $277.04
Catalog Number | Mfr. No. | Color | Thread Size | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Thread Size | Price | Quantity | ||||
03-340-319
|
DWK Life Sciences
73821C13 |
Red | 13 mm |
Case of 1000 Each for $164.06
|
|
03-340-322
|
DWK Life Sciences
73821C20 |
Red | 20 mm |
Case of 1000 Each for $179.28
|
|
03-340-321
|
DWK Life Sciences
73821B20 |
Blue | 20 mm |
Case of 1000 Each for $190.46
|
|
03-340-320
|
DWK Life Sciences
73821D13 |
Green | 13 mm |
Case of 1000 Each for $277.04
|
|
03-340-323
|
DWK Life Sciences
73821D20 |
Green | 20 mm |
Case of 1000 Each for $190.46
|
|
Description
- Allows total removal of seal without decappers
- For use with 13 and 20mm serum finished bottles and vials
- Designed to secure septa and stoppers to bottles and vials
Order Info
Available in blue, red, and green color to help separate and identify samples
Specifications
Tear-off | |
Aluminum | |
1000 Case |